Gene Transcripts

Gene Symbol
Bbs1
Entrez Gene ID
309156
Gene Name
Bardet-Biedl syndrome 1
Gene Aliases
Chromosome Location
Chr. 1: 220146247 - 220165545 on Build Rnor_6.0
UniGene ID
Rn.12497
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107569.1 NP_001101039.1 8 747
XM_008760137.1 XP_008758359.1 8 765

View Full Product Details