Gene Transcripts

Gene Symbol
Slitrk2
Entrez Gene ID
309349
Gene Name
SLIT and NTRK-like family, member 2
Gene Aliases
RGD1561361
Chromosome Location
Chr. X: 151099842 - 151111816 on Build Rnor_6.0
UniGene ID
Rn.106231
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107587.1 NP_001101057.1 2 1767
XM_006257625.2 XP_006257687.1 3 2049
XM_006257626.2 XP_006257688.1 4 1769
XM_008773586.1 XP_008771808.1 3 2060

View Full Product Details