Gene Transcripts

Gene Symbol
Myof
Entrez Gene ID
309499
Gene Name
myoferlin
Gene Aliases
Fer1l3, RGD1564216
Chromosome Location
Chr. 1: 256585410 - 256735054 on Build Rnor_6.0
UniGene ID
Rn.73651
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191636.1 NP_001178565.1 50 5760
XM_006231325.2 XP_006231387.1 51 6128
XM_006231326.2 XP_006231388.1 50 6089

View Full Product Details