Gene Transcripts

Gene Symbol
ZNF282
Entrez Gene ID
8427
Gene Name
zinc finger protein 282
Gene Aliases
HUB1
Chromosome Location
Chr. 7: 149195462 - 149226248 on Build GRCh38
UniGene ID
Hs.729056
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303481.1 NP_001290410.1 7 1275
NM_003575.3 NP_003566.1 7 1275
XM_006716151.3 XP_006716214.1 7 1272
GenBank mRNA AK299280.1 - 7 1187
AK316282.1 - 7 1162
AY927504.1 - 5 498
AY927505.1 - 6 948
BC041704.1 - 7 1002
BC055287.1 - 7 1065
BC073805.1 - 7 1168 6645861
D30612.1 - 7 1288

View Full Product Details