Gene Transcripts

Gene Symbol
Nol6
Entrez Gene ID
313167
Gene Name
nucleolar protein 6 (RNA-associated)
Gene Aliases
Chromosome Location
Chr. 5: 57444448 - 57473811 on Build Rnor_6.0
UniGene ID
Rn.224957
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006238112.2 XP_006238174.1 22 4046
XM_008763646.1 XP_008761868.1 22 4046
GenBank mRNA EF688596.1 - 10 1185

View Full Product Details