Gene Transcripts

Gene Symbol
Hs1bp3
Entrez Gene ID
313950
Gene Name
HCLS1 binding protein 3
Gene Aliases
RGD1311331
Chromosome Location
Chr. 6: 33517729 - 33548015 on Build Rnor_6.0
UniGene ID
Rn.23111
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_233975.8 XP_233975.5 4 617
GenBank mRNA FQ224595.1 - 4 586

View Full Product Details