Gene Transcripts

Gene Symbol
Baz1a
Entrez Gene ID
314126
Gene Name
bromodomain adjacent to zinc finger domain, 1A
Gene Aliases
Chromosome Location
Chr. 6: 75793223 - 75873854 on Build Rnor_6.0
UniGene ID
Rn.1864
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170568.1 NP_001164039.1 18 3159
GenBank mRNA CK595287.1 - 2 264

View Full Product Details