Gene Transcripts

Gene Symbol
Cdc42ep1
Entrez Gene ID
315121
Gene Name
CDC42 effector protein (Rho GTPase binding) 1
Gene Aliases
Chromosome Location
Chr. 7: 120067552 - 120071154 on Build Rnor_6.0
UniGene ID
Rn.164707
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079700.1 NP_001073168.1 2 659
GenBank mRNA BC128744.1 - 2 659

View Full Product Details