Gene Transcripts

Gene Symbol
Pih1d2
Entrez Gene ID
315645
Gene Name
PIH1 domain containing 2
Gene Aliases
RGD1310503
Chromosome Location
Chr. 8: 55050284 - 55061397 on Build Rnor_6.0
UniGene ID
Rn.105880
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191793.1 NP_001178722.1 6 1005
XM_006243028.2 XP_006243090.1 5 771
XM_006243029.2 XP_006243091.1 4 678
XM_006243030.2 XP_006243092.1 4 677

View Full Product Details