Gene Transcripts

Gene Symbol
A4gnt
Entrez Gene ID
685758
Gene Name
alpha-1,4-N-acetylglucosaminyltransferase
Gene Aliases
Chromosome Location
Chr. 8: 107856851 - 107866438 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766588.1 XP_008764810.1 3 935

View Full Product Details