Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001108216.1 | NP_001101686.1 | 4 | 683 | |
XM_006244825.2 | XP_006244887.1 | 4 | 648 | ||
XM_008767035.1 | XP_008765257.1 | 4 | 3092 | ||
GenBank mRNA | AY724492.1 | - | 4 | 683 |