Gene Transcripts

Gene Symbol
Bahcc1
Entrez Gene ID
360674
Gene Name
BAH domain and coiled-coil containing 1
Gene Aliases
RGD1306500
Chromosome Location
Chr. 10: 109424308 - 109482305 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008768494.1 XP_008766716.1 25 6658

View Full Product Details