Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003845.2 | NP_003836.1 | 6 | 662 | |
NR_104115.1 | 6 | 1038 | |||
GenBank mRNA | AF263541.1 | - | 5 | 562 | |
AK308260.1 | - | 8 | 883 | ||
BC031244.1 | - | 6 | 656 | 5266287 | |
GU139743.1 | - | 5 | 646 | ||
GU139746.1 | - | 6 | 1011 |