Gene Transcripts

Gene Symbol
Dak
Entrez Gene ID
361730
Gene Name
dihydroxyacetone kinase 2 homolog (S. cerevisiae)
Gene Aliases
RGD1311026
Chromosome Location
Chr. 1: 226642901 - 226657698 on Build Rnor_6.0
UniGene ID
Rn.55630
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039031.1 NP_001034120.1 9 995
XM_006231056.2 XP_006231118.1 9 881
XM_006231057.2 XP_006231119.2 9 1164
GenBank mRNA BC098925.1 - 9 995

View Full Product Details