Gene Transcripts
- Gene Symbol
- Clca4
- Entrez Gene ID
- 362053
- Gene Name
- chloride channel accessory 4
- Gene Aliases
- Clca6, Prp3
- Chromosome Location
- Chr. 2: 250843165 - 250864060 on Build Rnor_6.0
- UniGene ID
- Rn.44141
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_201419.1 |
NP_958822.1
|
10 |
1626 |
|
GenBank mRNA |
AY333961.1 |
- |
10 |
1626 |
|
View Full Product Details