Gene Transcripts
- Gene Symbol
- Hnrnpa3
- Entrez Gene ID
- 362152
- Gene Name
- heterogeneous nuclear ribonucleoprotein A3
- Gene Aliases
- Hnrpa3
- Chromosome Location
- Chr. 3: 62481228 - 62491356 on Build Rnor_6.0
- UniGene ID
- Rn.107690
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001111294.1 |
NP_001104764.1
|
4, 5 |
641 |
|
NM_001111295.1 |
NP_001104765.1
|
4, 5 |
575 |
|
NM_198132.3 |
NP_937765.1
|
4, 5 |
641 |
|
XM_008761911.1 |
XP_008760133.1
|
4, 5 |
590 |
|
XM_008761912.1 |
XP_008760134.1
|
4, 5 |
590 |
|
XM_008761913.1 |
XP_008760135.1
|
4, 5 |
636 |
|
GenBank mRNA |
AY363226.1 |
- |
4, 5 |
587 |
|
AY363227.1 |
- |
4, 5 |
521 |
|
BC081878.1 |
- |
4, 5 |
581 |
7108128 |
CB725005.1 |
- |
4, 5 |
513 |
|
FQ214617.1 |
- |
4, 5 |
537 |
|
View Full Product Details