Gene Transcripts

Gene Symbol
Dnajc24
Entrez Gene ID
362184
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 24
Gene Aliases
Dph4, RGD1564710, Zcsl3
Chromosome Location
Chr. 3: 96025388 - 96065744 on Build Rnor_6.0
UniGene ID
Rn.231483
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191853.1 NP_001178782.1 3 256
XM_006234653.2 XP_006234715.1 3 300
XM_006234654.2 XP_006234716.1 3 289

View Full Product Details