Gene Transcripts

Gene Symbol
Elmo2
Entrez Gene ID
362271
Gene Name
engulfment and cell motility 2
Gene Aliases
Chromosome Location
Chr. 3: 161812474 - 161849979 on Build Rnor_6.0
UniGene ID
Rn.2097
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134955.1 NP_001128427.1 21 2044
XM_006235599.2 XP_006235661.1 20 2020
GenBank mRNA BC089970.1 - 2 513

View Full Product Details