Gene Transcripts

Gene Symbol
ASAP2
Entrez Gene ID
8853
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 2
Gene Aliases
AMAP2, CENTB3, DDEF2, PAG3, PAP, Pap-alpha, SHAG1
Chromosome Location
Chr. 2: 9206765 - 9405683 on Build GRCh38
UniGene ID
Hs.555902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135191.1 NP_001128663.1 8, 9 1098
NM_003887.2 NP_003878.1 8, 9 1098
XM_006711898.1 XP_006711961.1 8, 9 851
XM_006711901.3 XP_006711964.1 8, 9 701
XM_006711902.1 XP_006711965.1 8, 9 851
XM_011510403.2 XP_011508705.1 8, 9 872
XM_011510404.2 XP_011508706.1 8, 9 871
XM_011510405.2 XP_011508707.1 7, 8 798
XM_011510406.2 XP_011508708.1 9, 10 805
XM_011510407.2 XP_011508709.1 8, 9 872
XM_011510408.2 XP_011508710.1 8, 9 875
XM_011510409.1 XP_011508711.1 8, 9 743
GenBank mRNA AB007860.1 - 8, 9 1098
BC063308.1 - 8, 9 1051 30345039

View Full Product Details