Gene Transcripts

Gene Symbol
ASAP2
Entrez Gene ID
8853
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 2
Gene Aliases
AMAP2, CENTB3, DDEF2, PAG3, PAP, Pap-alpha, SHAG1
Chromosome Location
Chr. 2: 9206765 - 9405683 on Build GRCh38
UniGene ID
Hs.555902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135191.1 NP_001128663.1 17 2054
NM_003887.2 NP_003878.1 17 2054
XM_006711898.1 XP_006711961.1 18 1816
XM_006711901.3 XP_006711964.1 18 1666
XM_006711902.1 XP_006711965.1 18 1816
XM_011510403.2 XP_011508705.1 18 1837
XM_011510404.2 XP_011508706.1 17 1827
XM_011510405.2 XP_011508707.1 17 1763
XM_011510406.2 XP_011508708.1 19 1770
XM_011510407.2 XP_011508709.1 18 1837
XM_011510408.2 XP_011508710.1 18 1840
XM_011510409.1 XP_011508711.1 18 1708
GenBank mRNA AB007860.1 - 17 2054
BC063308.1 - 17 2007 30345039

View Full Product Details