Gene Transcripts
- Gene Symbol
- Srsf7
- Entrez Gene ID
- 362687
- Gene Name
- serine/arginine-rich splicing factor 7
- Gene Aliases
- Sfrs7
- Chromosome Location
- Chr. 6: 2879312 - 2886505 on Build Rnor_6.0
- UniGene ID
- Rn.218757
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039035.2 |
NP_001034124.2
|
2, 3 |
313 |
|
XM_006239642.1 |
XP_006239704.1
|
2, 3 |
353 |
|
XM_006239643.1 |
XP_006239705.1
|
2, 3 |
353 |
|
XM_006239644.1 |
XP_006239706.1
|
2, 3 |
353 |
|
XM_006239645.1 |
XP_006239707.1
|
2, 3 |
353 |
|
XM_006239646.1 |
XP_006239708.1
|
2, 3 |
353 |
|
GenBank mRNA |
BC099175.1 |
- |
2, 3 |
303 |
|
FM084661.1 |
- |
2, 3 |
313 |
|
FM085512.1 |
- |
2, 3 |
307 |
|
FQ219847.1 |
- |
2, 3 |
319 |
|
FQ229134.1 |
- |
2, 3 |
320 |
|
View Full Product Details