Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006773.3 | NP_006764.3 | 3 | 530 | |
GenBank mRNA | AK001467.1 | - | 3 | 502 | |
BC001238.1 | - | 3 | 459 | 3452935 | |
BC003360.1 | - | 3 | 489 | 2900252 | |
BC024739.1 | - | 3 | 504 | 4995871 | |
CR457060.1 | - | 2 | 251 | ||
X98743.1 | - | 2 | 268 |