Gene Transcripts

Gene Symbol
Hdac10
Entrez Gene ID
362981
Gene Name
histone deacetylase 10
Gene Aliases
Chromosome Location
Chr. 7: 130102335 - 130108346 on Build Rnor_6.0
UniGene ID
Rn.107028
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035000.1 NP_001030172.1 11, 12 1197
XM_006242220.1 XP_006242282.1 11, 12 2106
GenBank mRNA BC092573.1 - 11, 12 1197

View Full Product Details