Gene Transcripts

Gene Symbol
HSPB3
Entrez Gene ID
8988
Gene Name
heat shock protein family B (small) member 3
Gene Aliases
DHMN2C, HMN2C, HSPL27
Chromosome Location
Chr. 5: 54455601 - 54456384 on Build GRCh38
UniGene ID
Hs.41707
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006308.2 NP_006299.1 1 287
GenBank mRNA AK314215.1 - 1 288
BC126272.1 - 1 142 8991988
BC132869.1 - 1 154 40146891
BX106905.1 - 1 272
CR450314.1 - 1 98
U15590.1 - 1 273
Y17782.1 - 1 246

View Full Product Details