Gene Transcripts

Gene Symbol
Micb
Entrez Gene ID
365212
Gene Name
MHC class I polypeptide-related sequence B
Gene Aliases
Mill2
Chromosome Location
Chr. 1: 79754587 - 79772092 on Build Rnor_6.0
UniGene ID
Rn.215725
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017468.2 NP_001017468.1 4 888
GenBank mRNA AB113963.1 - 4 878

View Full Product Details