Gene Transcripts

Gene Symbol
Olr63
Entrez Gene ID
365323
Gene Name
olfactory receptor 63
Gene Aliases
POGR, ratchr1-160727936-160726983_ORF
Chromosome Location
Chr. 1: 167864306 - 167873267 on Build Rnor_6.0
UniGene ID
Rn.129398
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000542.1 NP_001000542.1 1 112
XM_006229890.2 XP_006229952.1 2 429
GenBank mRNA AY834218.1 - 1 112

View Full Product Details