Gene Transcripts

Gene Symbol
Selt
Entrez Gene ID
365802
Gene Name
selenoprotein T
Gene Aliases
Chromosome Location
Chr. 2: 148765412 - 148781080 on Build Rnor_6.0
UniGene ID
Rn.18892
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014253.2 NP_001014275.2 3, 4 429
GenBank mRNA AY995234.1 - 3, 4 429
BC086953.1 - 3, 4 323 7321890
FQ211813.1 - 447

View Full Product Details