Gene Transcripts

Gene Symbol
Bola1
Entrez Gene ID
365875
Gene Name
bolA family member 1
Gene Aliases
RGD1310586
Chromosome Location
Chr. 2: 198338114 - 198339130 on Build Rnor_6.0
UniGene ID
Rn.13864
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001071776.1 NP_001065244.1 2 419
GenBank mRNA DQ912022.1 - 2 419

View Full Product Details