Gene Transcripts

Gene Symbol
Rbm38
Entrez Gene ID
366262
Gene Name
RNA binding motif protein 38
Gene Aliases
Rnpc1
Chromosome Location
Chr. 3: 171037896 - 171050500 on Build Rnor_6.0
UniGene ID
Rn.17628
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108965.1 NP_001102435.1 3 1222
XM_006235704.2 XP_006235766.1 4 692
XM_008762528.1 XP_008760750.1 5 782

View Full Product Details