Gene Transcripts

Gene Symbol
Lrg1
Entrez Gene ID
367455
Gene Name
leucine-rich alpha-2-glycoprotein 1
Gene Aliases
Chromosome Location
Chr. 9: 10952424 - 10954664 on Build Rnor_6.0
UniGene ID
Rn.17847
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009717.1 NP_001009717.1 2 88
GenBank mRNA BC088434.1 - 2 88
FQ220477.1 - 2 98

View Full Product Details