Gene Transcripts

Gene Symbol
Bcl7b
Entrez Gene ID
368001
Gene Name
B-cell CLL/lymphoma 7B
Gene Aliases
Chromosome Location
Chr. 12: 24543748 - 24557093 on Build Rnor_6.0
UniGene ID
Rn.37635
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109021.1 NP_001102491.1 5 689
GenBank mRNA BC160857.1 - 5 572

View Full Product Details