Gene Transcripts

Gene Symbol
Olr36
Entrez Gene ID
405257
Gene Name
olfactory receptor 36
Gene Aliases
Chromosome Location
Chr. 1: 164717029 - 164717937 on Build Rnor_6.0
UniGene ID
Rn.141937
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000935.1 NP_001000935.1 1 228

View Full Product Details