Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004720.5 | NP_004711.2 | 3 | 1104 | |
XM_011528421.2 | XP_011526723.1 | 3 | 1468 | ||
XM_017027468.1 | XP_016882957.1 | 5 | 1399 | ||
XM_017027469.1 | XP_016882958.1 | 4 | 1202 | ||
GenBank mRNA | AF011466.1 | - | 3 | 1049 | |
AF197929.1 | - | 3 | 1083 | ||
AF233092.1 | - | 2 | 971 | ||
AK123043.1 | - | 6 | 1868 | ||
AK316573.1 | - | 3 | 1065 | ||
AY322548.1 | - | 2 | 965 | ||
BC025695.1 | - | 3 | 1144 | 5209267 |