Gene Transcripts

Gene Symbol
Ctsql2
Entrez Gene ID
408201
Gene Name
cathepsin Q-like 2
Gene Aliases
Chromosome Location
Chr. 17: 4062294 - 4068108 on Build Rnor_6.0
UniGene ID
Rn.128678
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002813.2 NP_001002813.2 5 709
XM_008771444.1 XP_008769666.1 5 693
GenBank mRNA BC097257.1 - 5 709

View Full Product Details