Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256910.1 | NP_001243839.1 | 14 | 2287 | |
NM_004728.3 | NP_004719.2 | 14 | 2168 | ||
XM_011540336.1 | XP_011538638.1 | 13 | 1998 | ||
XM_017016910.1 | XP_016872399.1 | 13 | 1916 | ||
GenBank mRNA | AB062394.1 | - | 6 | 647 | |
AK025629.1 | - | 11 | 1470 | ||
AK315585.1 | - | 14 | 2117 | ||
BC008071.2 | - | 14 | 2106 | 2987980 | |
BX648405.1 | - | 14 | 2054 | ||
CR749598.1 | - | 14 | 2288 | ||
U41387.1 | - | 14 | 2119 |