Gene Transcripts

Gene Symbol
Vom1r6
Entrez Gene ID
494231
Gene Name
vomeronasal 1 receptor 6
Gene Aliases
V1re2
Chromosome Location
Chr. 1: 60044985 - 60045881 on Build Rnor_6.0
UniGene ID
Rn.129459
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009512.1 NP_001009512.1 1 279
GenBank mRNA AY510290.1 - 1 279

View Full Product Details