Gene Transcripts

Gene Symbol
Vom1r82
Entrez Gene ID
494235
Gene Name
vomeronasal 1 receptor 82
Gene Aliases
V1rc41
Chromosome Location
Chr. 4: 88119838 - 88120734 on Build Rnor_6.0
UniGene ID
Rn.129658
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009515.1 NP_001009515.1 1 482
GenBank mRNA AY510313.1 - 1 482

View Full Product Details