Gene Transcripts

Gene Symbol
Fn3k
Entrez Gene ID
498034
Gene Name
fructosamine 3 kinase
Gene Aliases
Fnsk
Chromosome Location
Chr. 10: 110626473 - 110642262 on Build Rnor_6.0
UniGene ID
Rn.233745
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109051.1 NP_001102521.1 4 364
XM_006247907.2 XP_006247969.1 6 1260
GenBank mRNA FQ212712.1 - 8 825

View Full Product Details