Gene Transcripts

Gene Symbol
RAB11B
Entrez Gene ID
9230
Gene Name
RAB11B, member RAS oncogene family
Gene Aliases
H-YPT3
Chromosome Location
Chr. 19: 8390321 - 8404434 on Build GRCh38
UniGene ID
Hs.558403
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004218.3 NP_004209.2 4 594
GenBank mRNA AF498947.1 - 4 498
AK225036.1 - 4 512
AK225642.1 - 4 514
AK297498.1 - 4 522
AK310573.1 - 4 285
AK312994.1 - 4 548
BC110081.1 - 4 500 40035112
BT019535.1 - 4 498
BT019536.1 - 4 498
BX647356.1 - 2596
CR536494.1 - 4 498
CR541691.1 - 4 498
CX787570.1 - 2 145
D38516.1 - 4 507
EF560724.1 - 4 577
X79780.1 - 4 504

View Full Product Details