Gene Transcripts

Gene Symbol
Spdye4
Entrez Gene ID
498146
Gene Name
speedy/RINGO cell cycle regulator family member E4
Gene Aliases
Chromosome Location
Chr. 12: 13390859 - 13398301 on Build Rnor_6.0
UniGene ID
Rn.137830
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034153.1 NP_001029325.1 1 221
GenBank mRNA BC105855.1 - 1 221

View Full Product Details