Gene Transcripts

Gene Symbol
Thnsl1
Entrez Gene ID
498805
Gene Name
threonine synthase-like 1 (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 17: 88095830 - 88102264 on Build Rnor_6.0
UniGene ID
Rn.198366
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025035.1 NP_001020206.1 4 680
GenBank mRNA BC091213.1 - 4 680

View Full Product Details