Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001047111.2 | NP_001040576.1 | 2 | 416 | |
XM_006235610.1 | XP_006235672.1 | 3 | 479 | ||
XM_006235612.1 | XP_006235674.1 | 3 | 235 | ||
XM_006235614.2 | XP_006235676.1 | 3 | 378 | ||
XM_006235615.2 | XP_006235677.1 | 3 | 324 | ||
GenBank mRNA | AY113696.1 | - | 2 | 294 | |
BC158561.1 | - | 2 | 416 | 7387488 | |
FQ213987.1 | - | 2 | 328 | ||
FQ220039.1 | - | 3 | 304 |