Gene Transcripts

Gene Symbol
Kbtbd8
Entrez Gene ID
500262
Gene Name
kelch repeat and BTB (POZ) domain containing 8
Gene Aliases
RGD1563166
Chromosome Location
Chr. 4: 127164738 - 127177537 on Build Rnor_6.0
UniGene ID
Rn.219562
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109250.3 NP_001102720.2 3 488
XM_006236932.1 XP_006236994.1 3 485
GenBank mRNA BC160902.1 - 3 488

View Full Product Details