Gene Transcripts

Gene Symbol
Rbm44
Entrez Gene ID
501183
Gene Name
RNA binding motif protein 44
Gene Aliases
RGD1566074
Chromosome Location
Chr. 9: 98279033 - 98305248 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008767346.1 XP_008765568.1 3, 4 1722
XM_576610.6 XP_576610.2 3, 4 1863

View Full Product Details