Gene Transcripts

Gene Symbol
CDY2A
Entrez Gene ID
9426
Gene Name
chromodomain protein, Y-linked, 2A
Gene Aliases
CDY, CDY2
Chromosome Location
Chr. Y: 18025787 - 18027746 on Build GRCh38
UniGene ID
Hs.251375
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004825.2 NP_004816.1 1 1395
GenBank mRNA AF080598.1 - 1 1395
AK292233.1 - 1 1368

Gene Symbol
CDY2B
Entrez Gene ID
203611
Gene Name
chromodomain protein, Y-linked, 2B
Gene Aliases
CDY
Chromosome Location
Chr. Y: 17878260 - 17880219 on Build GRCh38
UniGene ID
Hs.532657
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001722.1 NP_001001722.1 1 1394
GenBank mRNA BC069087.1 - 1 1394
BC130426.1 - 1 1163 40146457
BC130428.1 - 1 1163 40146459

View Full Product Details