Gene Transcripts
- Gene Symbol
- Cenpv
- Entrez Gene ID
- 501702
- Gene Name
- centromere protein V
- Gene Aliases
- Prr6, RGD1565681
- Chromosome Location
- Chr. 10: 48846513 - 48861895 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_577104.6 |
XP_577104.3
|
5 |
1586 |
|
View Full Product Details