Gene Transcripts

Gene Symbol
Mrgprb2
Entrez Gene ID
502338
Gene Name
MAS-related GPR, member B2
Gene Aliases
Mgrg10, MrgB6, Mrgprb6
Chromosome Location
Chr. 1: 104022906 - 104024711 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_577804.4 XP_577804.2 1 107

View Full Product Details