Gene Transcripts

Gene Symbol
Eif2a
Entrez Gene ID
502531
Gene Name
eukaryotic translation initiation factor 2A
Gene Aliases
Chromosome Location
Chr. 2: 148722347 - 148755781 on Build Rnor_6.0
UniGene ID
Rn.198593
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109339.1 NP_001102809.1 10 798
XM_006232388.2 XP_006232450.1 10 945
GenBank mRNA FQ226213.1 - 3 247

View Full Product Details