Gene Transcripts

Gene Symbol
Rerg
Entrez Gene ID
502916
Gene Name
RAS-like, estrogen-regulated, growth-inhibitor
Gene Aliases
RGD1562829
Chromosome Location
Chr. 4: 171069161 - 171176678 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006237582.2 XP_006237644.1 5 710
XM_578417.6 XP_578417.4 5 715

View Full Product Details