Gene Transcripts

Gene Symbol
ADAMTS3
Entrez Gene ID
9508
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 3
Gene Aliases
ADAMTS-4
Chromosome Location
Chr. 4: 72280969 - 72568799 on Build GRCh38
UniGene ID
Hs.590919
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014243.2 NP_055058.2 4 632
XM_011532421.2 XP_011530723.1 4 664
XM_011532422.2 XP_011530724.1 4 1163
GenBank mRNA AB002364.1 - 4 584
AF247668.1 - 4 632
BC130287.1 - 4 629 40146318
BC132735.1 - 4 629 40146757

View Full Product Details